Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Flunitrazepam-13C6, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  Intermetro Industries Corp 1836NFST316

Catalog No. 01-001-362


Only null left
Add to Cart

Description

Description

This stable-labeled internal standard is suitable for quantitation of flunitrazepam levels by LC-MS/MS methods for clinical toxicology, forensic analysis, urine drug testing, pain prescription monitoring or isotope dilution applications. Flunitrazepam is an intermediate-acting benzodiazepine marketed under the trade name Rohypnol as a hypnotic, sedative, anticonvulsant and skeletal muscle relaxant. For these analyses, 13C-labeled internal standards can eliminate potential issues of isotopic exchange and minimize separation of later-eluting analytes from a higher substituted deuterium-labeled internal standard.
Specifications

Specifications

67-56-1, 1622-62-4
MFCD00057906
100 μg/mL in methanol
Ampule
C1013C6H12FN3O3
UN 1230 - class 3 - PG 2 - Methanol, solution
319.24
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

CERILLIANT is a registered trademark of Cerilliant Corporation