Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Glutethimide Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  Intermetro Industries Corp 2430NFST316

Catalog No. 01-001-401


Only null left
Add to Cart

Description

Description

Glutethimide is a hypnotic sedative sold under the trade name Doriden. Glutethimide has a history of abuse in combination with codeine. This Certified Snap-N-Spike™ Solution is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from urine drug testing to forensic analysis and clinical toxicology.
Specifications

Specifications

67-56-1, 77-21-4
UN 1230 - class 3 - PG 2 - Methanol, solution
217.26
1 mL
C13H15NO2
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

CERILLIANT is a registered trademark of Cerilliant Corporation