Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Hydroxy Iloperidone Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  Intermetro Industries Corp 2448NFST316

Catalog No. 01-001-453


Only null left
Add to Cart

Description

Description

A certified Snap-N-Spike™ solution suitable for hydroxy iloperidone testing methods by GC/MS or LC/MS for clinical toxicology, forensic testing, or pharmaceutical research. Hydroxy iloperidone is a major metabolite of the atypical antipsychotic iloperidone which is administered for the treatment of schizophrenia. The metabolite is also pharmacologically active, possessing central nervous system (CNS) activity.
Specifications

Specifications

67-56-1, 133454-55-4
UN 1230 - class 3 - PG 2 - Methanol
428.50
1 mL
C24H29FN2O4
100 μg/mL in methanol
Ampule
9°C (48.2°F)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

CERILLIANT is a registered trademark of Cerilliant Corporation