Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ L-Thyroxine (T4) Solution, 100 μg/mL in Methanol with 0.1N NH3, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T0731ML

Catalog No. 01-002-070


Add to Cart

Description

Description

Thyroxine or T4 is a thyroid hormone responsible for regulation of the body's basal metabolic rate and essential for growth, development and heart and nervous system function. Free and total T4 serum levels are measured by LC-MS/MS for determination of hyper- or hypothyroidism as well as for monitoring treatment with synthetic thyroid hormones. This Certified Spiking Solution™ is suitable for use in LC-MS/MS testing methods as starting materials in preparation of linearity standards, calibrators, and controls.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 51-48-9
HOC6H2(I)2OC6H2(I)2CH2CH(NH2)CO2H
UN 1230 - class 3 - PG 2 - Methanol, solution
L-Thyroxine, 3,3′,5,5′′-Tetraiodo-L-thyronine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine, T4
Ampule
9.7°C (49.5°F) - closed cup
223°C (dec.) (lit.)
MFCD00002595
100 μg/mL in methanol with 0.1N NH3
776.87
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation