Learn More
CERILLIANT™ Lormetazepam-13C,D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco L0511ML
Catalog No. 01-001-555
Description
Description
Lormetazepam is a benzodiazepine with sedative-hypnotic, anxiolytic, anticonvulsant, and muscle relaxant effects. Lormetazepam is sold under trade names such as Loramet and Noctamid for treatment of insomnia. This Snap-N-Shoot™ stable-labeled internal standard is suitable for quantitation of lormetazepam levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, or forensic analysis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
67-56-1 , 1285932-00-4
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C1513CH9D3Cl2N2O2
339.20
1 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Title
Select an issue
CERILLIANT is a registered trademark of Cerilliant Corporation