Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Temazepam Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T9071ML

Catalog No. 01-002-117


Add to Cart

Description

Description

Sold under the trade names Restoril™ and Normison, temazepam is a benzodiazepine approved for the short term treatment of insomnia. The drug has anti-anxiety, anticonvulsant and skeletal muscle relaxant properties. This certified Snap-N-Spike™ solution is suitable for a variety of GC/MS and LC/MS testing applications from forensic analysis and pain prescription monitoring to clinical toxicology and urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
MFCD00057904
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C16H13ClN2O2
UN 1230 - class 3 - PG 2 - Methanol, solution
300.74
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation