Learn More
Enzo Life Sciences Ceritinib (free base) (25mg). CAS: 1032900-25-6
SDP
Supplier:  Enzo Life Sciences ENZCHM2180025
Encompass_Preferred
Ceritinib is a selective inhibitor of ALK, a target found in metastatic non-small cell lung cancer. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3564
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.