Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) chloride heptahydrate, 99%

Catalog No. p-222781
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211820250 25 g Glass bottle
AC211821000 100 g Glass bottle
AC211825000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC211820250 Supplier Thermo Scientific Chemicals Supplier No. 211820250
Only null left
Add to Cart

CAS: 18618-55-8 | CeCl3·7H2O | 372.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18618-55-8
Molecular Formula CeCl3·7H2O
Molecular Weight (g/mol) 372.58
MDL Number MFCD00149634

Specifications

Melting Point 848.0°C
Color Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula CeCl3·7H2O
Fieser 01,121; 09,99; 15,72; 16,4; 17,314
Merck Index 10, 1956
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 372.58
Percent Purity 99%
Packaging Glass bottle
Density 3.97
Chemical Name or Material Cerium(III) chloride heptahydrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.