Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) iodide, ultra dry, 99.9% (REO)

Catalog No. AA1364106
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1364106 5 g
AA1364103 1 g
AA1364114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1364106 Supplier Thermo Scientific Chemicals Supplier No. 01364106
Add to Cart

Used as a pharmaceutical intermediate. | CAS: 7790-87-6 | CeI3

Cerium(III) iodide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-87-6
Molecular Formula CeI3
MDL Number MFCD00049423
Synonym Cerium triiodide

Specifications

Melting Point 766°C
Packaging Ampouled under argon
Quantity 5 g
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water
Formula Weight 520.83
Percent Purity 99.9%
Physical Form Powder
Chemical Name or Material Cerium(III) iodide
Hazard Category H317-H334-H360
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 232-228-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.