Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) nitrate hexahydrate, 99.5%

Catalog No. ac218691000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218691000 100 g Plastic bottle
AC218695000 500 g Plastic bottle
AC218690025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC218691000 Supplier Thermo Scientific Chemicals Supplier No. 218691000
Only null left
Add to Cart

CAS: 10294-41-4 | CeH12N3O15 | 434.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-41-4
Molecular Formula CeH12N3O15
Molecular Weight (g/mol) 434.22
MDL Number MFCD00149631
InChI Key QQZMWMKOWKGPQY-UHFFFAOYSA-N
Synonym cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5f
PubChem CID 16211466
SMILES O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Colorless to White
Physical Form Crystalline Powder or Agglomerating Crystals
Quantity 100 g
Assay Percent Range 99.4% min. (CeO2/TREO basis)
Linear Formula Ce(NO3)3·6H2O
Fieser 16,67
Merck Index 10,1959
Solubility Information Solubility in water: 1754g/L (25°C). Other solubilities: soluble in alcohol and acetone
IUPAC Name cerium(3+) hexahydrate trinitrate
Formula Weight 434.22
Percent Purity 99.5%
Packaging Plastic bottle
Chemical Name or Material Cerium(III) nitrate hexahydrate, 99.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
In case of major
RTECSNumber FK6300000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.