Learn More
Cerium(IV) and Dilute Nitric acid, Etchant Solution, Ce(IV) concentration 0.25N, Thermo Scientific Chemicals
Description
It is widely applied as etchant solution in industrial purpose to chemically remove layers from the surface of a wafer during manufacturing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 1 L |
| UN Number | UN3098 |
| Solubility Information | Soluble in water |
| Concentration or Composition (by Analyte or Components) | Ce(IV) concentration 0.25N |
| Chemical Name or Material | Cerium(IV) and Dilute Nitric acid, Etchant Solution |
Safety and Handling
| Hazard Category | H290-H314-H317-H335 |
| Hazard Statement | GHS H Statement H272-H315-H319 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P234-P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: III |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.