Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(IV) fluoride hydrate, Thermo Scientific Chemicals

Catalog No. p-7050544
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3858009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3858009 Supplier Thermo Scientific Chemicals Supplier No. 03858009
Only null left
Add to Cart

Cerium(IV) fluoride hydrates a water insoluble Cerium source for use in oxygen-sensitive applications, such as metal production. Commercial applications for cerium include metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. In steel manufacturing it is used to remove free oxygen and sulfur by forming stable oxysulfides and by tying up undesirable trace elements, such as lead and antimony.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60627-09-0
Molecular Formula CeF4
Molecular Weight (g/mol) 216.11
MDL Number MFCD00049422
InChI Key SWURHZJFFJEBEE-UHFFFAOYSA-J
Synonym cerium iv fluoride hydrate, acmc-20alhu, cerium 4+ fluoride-water 1/4/1
PubChem CID 71317457
IUPAC Name cerium(4+);tetrafluoride;hydrate
SMILES F[Ce](F)(F)F

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN3288
Solubility Information Insoluble in water; Slowly hydrolyzed by cold water
Formula Weight 216.11 (Anhydrous)
Chemical Name or Material Cerium(IV) fluoride hydrate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.