Learn More
Invitrogen™ CERK Polyclonal Antibody

Description
A suggested positive control for this product is heart, brain, skeletal muscle, kidney or liver lysate.
Specifications
Specifications
| Antigen | CERK |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | CERK |
| Gene Accession No. | Q8TCT0 |
| Gene Alias | acylsphingosine kinase; AI848610; Ceramide kinase; Cerk; CERK antibody; D330016D08Rik; dA59H18.2; dA59H18.3; hCERK; KIAA1646; Lipid kinase 4; lipid kinase LK4; LK4; mCERK; mKIAA1646 |
| Gene Symbols | CERK |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.