Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7211766 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5043 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5043 Supplier Proteintech Group Inc Supplier No. 257311AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CERK has 2 isoforms with the molecular weight of 60 and 38 kDa.

CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).

Specifications

Antigen CERK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CERK
Gene Accession No. Q8K4Q7, Q8TCT0
Gene Alias Acylsphingosine kinase, ceramide kinase, CERK, dA59H18.2, dA59H18.3, hCERK, KIAA1646, Lipid kinase 4, LK4
Gene Symbols CERK
Host Species Rabbit
Immunogen CERK Fusion Protein Ag22774
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223753, 64781
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.