Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ CERTAN™ Capillary Bottles

Catalog No. 111010641 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
1.5 mL
4.5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-101-0641 10 mL
11-100-4345 1.5 mL
11-101-0857 4.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-0641 Supplier MilliporeSigma™ Supelco™ Supplier No. 444215EA
Add to Cart
Add to Cart

To extract a solution from CERTAN™ Capillary Vials use a syringe with a needle diameter of 0.8 mm or smaller, and a minimum length of 70 mm

  • Unique, screw-cap sample containers
  • Designed for the storage of volatile samples and reference standards
  • Vials provide the advantage of a sealed ampule with the flexibility of a screw cap bottle or a septum vial
  • No appreciable change in concentration, even when vial is opened
  • Sample filling and removal using a standard GC syringe
  • Capillary Opening (D x H): 1.2 × 28 mm
  • Secure storage with minimal evaporation
  • Minimizes contamination risk
  • Essentially spillproof
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) PTFE
Diameter (Metric) Opening 1.2 mm
Color Clear
Diameter (Metric) Outer 30 mm
Material Glass
Volume (Metric) 10 mL
For Use With (Application) Storing Volatile Samples and Reference Standards
Graduated No
Closure Type Screw Cap
Quantity 5/Pk.
Height (Metric) 71 mm
Product Type Capillary Vial
Show More Show Less

CERTAN is a registered trademark of LGC Group

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.