Learn More
Certified Safety Certi-Strip™ Woven Adhesive Bandages

Description
- Woven Certi-Strips stretch for more comfort
- Adhesive that sticks and won't hurt when removed
- Medium weight with extra-strength adhesive
- Comes in standard sizes, as well as sizes for fingertip and knuckle
- Latex free
- Blue metal detector strips also available
Blue Medical Detector Strips (BMD)
- Flexible, heavy-duty fabric that is easily visible in case of loss
- Foil strip makes detection possible with most metal detectors
Specifications
Specifications
| For Use With (Application) | Protects minor cuts and scrapes |
| Product Type | Certi-strip Woven Bandage |
| Color | Tan |
| Material | Fabric |
| Sterility | Sterile |
| Quantity | 8 Pk. |
| Style | Knuckle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.