Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Certified Clean Borosilicate Glass Vials

Catalog No. 111003459 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
No. per Pack:
6
100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
11-100-3459 100
11-100-5112 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-3459 Supplier MilliporeSigma™ Supelco™ Supplier No. ULTRA100100EA
Add to Cart
Add to Cart

Borosilicate glass vials with certificate included

  • Case includes individual certificate
  • Certified Vials have <0.010 mg/L C from TOC
  • Include dust caps
  • Cleaning Products Cosmetics
  • Environmental Flavors and Fragrances
  • Food and Beverages
  • Genteral Analytical Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Cap 24 mm
Material Silicone, PTFE
For Use With (Application) TOC Analysis
No. per Pack 100
Volume (English) 40 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.