Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Certified Microcentrifuge Tubes in Self-Standing Bags

Catalog No. 220101221 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Amber
Blue
Green
Natural
Orange
Red
Violet
Yellow
Volume (Metric):
0.5mL
1.5mL
2mL
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric)
22-010-1221 Yellow 2mL
22-010-090 Natural 1.5mL
22-010-1199 Natural 0.5mL
22-010-1200 Amber 0.5mL
22-010-1201 Blue 0.5mL
22-010-1202 Green 0.5mL
22-010-1203 Orange 0.5mL
22-010-1204 Red 0.5mL
22-010-1205 Violet 0.5mL
22-010-1206 Yellow 0.5mL
22-010-1207 Amber 1.5mL
22-010-1208 Blue 1.5mL
22-010-1209 Green 1.5mL
22-010-1210 Orange 1.5mL
22-010-1211 Red 1.5mL
22-010-1212 Violet 1.5mL
22-010-1213 Yellow 1.5mL
22-010-1214 Natural 2mL
22-010-1215 Amber 2mL
22-010-1216 Blue 2mL
22-010-1217 Green 2mL
22-010-1218 Orange 2mL
22-010-1219 Red 2mL
22-010-1220 Violet 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. 22-010-1221 Supplier Globe Scientific Supplier No. 111574Y
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Globe Scientific Certified Microcentrifuge Tubes in Self-Standing Bags are resistant to most chemical reagents and can be autoclaved at 121°C.

  • Manufactured from FDA approved resins
  • Certified RNase, DNase, DNA and Pyrogen free
  • Supplied in resealable, tamper-evident self-standing bags
  • Large, clearly printed graduations
  • Frosted writing area on each tube
  • Smooth surfaces won't catch or tear gloves
  • Thin cap membrane for trouble-free needle piercing
  • Fit all standard and high capacity centrifuge rotors
  • Can be centrifuged at up to 20,000 RCF
  • Withstand temperatures from −80°C to +121°C
  • Food grade

Specifications

Certifications/Compliance RNase, DNase, DNA and Pyrogen Free
Autoclavable Autoclavable
Capacity (Metric) 2.0 mL
Color Yellow
Material Polypropylene
No. per Pack 500
Packaging Self-standing tamper evident bag
Volume (Metric) 2mL
Sterility Non-sterile
Graduated Yes
Closure Type Snap Cap
Max. RCF 20,000 x g
Product Type Microcentrifuge Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.