Learn More
Apexbio Technology LLC CERULETIDE 5MG
SDP
Supplier:  Apexbio Technology LLC B84655
Encompass_Preferred
Ceruletide (CAS 17650-98-5) is a synthetic decapeptide structurally and functionally analogous to the gastrointestinal hormone cholecystokinin (CCK) It acts primarily via binding to CCK receptors stimulating gastric pancreatic and biliary secretions as well as inducing contraction in gastrointestinal smooth muscle tissues In biomedical research ceruletide serves as a valuable tool for investigating pancreatic function digestive physiology and related gastrointestinal disorders
Catalog No. 50-224-0019
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.