Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceruloplasmin Mouse anti-Human, Rat, Clone: 3C8A5, Proteintech
SDP

Catalog No. p-7213493 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6590 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6590 Supplier Proteintech Group Inc Supplier No. 661561IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe (II)transferrin to Fe (III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.

Specifications

Antigen Ceruloplasmin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3C8A5
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CP
Gene Accession No. P00450, P13635
Gene Alias Ceruloplasmin, ceruloplasmin (ferroxidase), CP, CP 2, Ferroxidase
Gene Symbols CP
Host Species Mouse
Immunogen Ceruloplasmin Fusion Protein Ag15535
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1356, 24268
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.