Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium (8,9,12,8',9',12'-hexabromo-1,2-dicarbollide)cobaltate, Thermo Scientific™

Catalog No. AA4475006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4475006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4475006 Supplier Thermo Scientific Chemicals Supplier No. 4475006

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C4H16B18Br6CsCo
MDL Number MFCD08064213

Specifications

Color Brown
Linear Formula Cs[(C2B9Br3H8)2Co]
Quantity 5 g
Formula Weight 930.01
Physical Form Powder
Chemical Name or Material Cesium (8,9,12,8',9',12'-hexachloro-1,2-dicarbollide)cobaltate
Hazard Category Warning
Hazard Statement GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.