Learn More
Cesium (8,9,12,8',9',12'-hexachloro-1,2-dicarbollide)cobaltate, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | Brown |
| Linear Formula | Cs[(C2B9Cl3H8)2Co] |
| Quantity | 5 g |
| Formula Weight | 663.31 |
| Physical Form | Powder |
| Chemical Name or Material | Cesium (8,9,12,8',9',12'-hexachloro-1,2-dicarbollide)cobaltate |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H341-H317 Suspected of causing genetic defects. May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement P261-P280-P281-P363-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. Use personal protective equipment as required. Wash contaminated clothing before reuse. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.