Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Chloride, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1020380025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1020380025 25 g Glass bottle
M1020380100 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1020380025 Supplier MilliporeSigma Supplier No. 1020380025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7647-17-8 | ClCs | 168.36 g/mol

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Melting Point 646°C
pH 7 to 9
Physical Form Solid
Quantity 25 g
Assay Percent Range ≥99.5% (argentometric)
Solubility Information 1860 g/L in Water
InChI Formula 1S/ClH.Cs/h1H;/q;+1/p-1
Formula Weight 168.36
Grade Analytical
Concentration or Composition (by Analyte or Components) Al- ≤0.0005%, Cu- ≤0.0003%, Fe- ≤0.0003%, K- ≤0.002%, Li- ≤0.00005%, Mg- leq;0.0005%, N- ≤0.001%, Na- ≤0.002%, Pb- ≤0.0001%, Rb- ≤0.008%, Zn- ≤0.0003%
Odor Odorless
Packaging Glass bottle
Product Line EMSURE™
Residue after Ignition ≤0.002%
Sensitivity Hygroscopic
Appearance White solid
Decomposition Information Hydrogen Chloride, Cesium Oxides
Density 3.97 g/cm³
Chemical Name or Material Cesium chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.