Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium chloride, Free-flowing, Reagent Grade, anhydrous, 99.9%, Solstice

Catalog No. p-7111976 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-524 1 kg Poly Bottle
60-047-525 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-524 Supplier Solstice Supplier No. 7464871KG
Only null left
Add to Cart
Add to Cart

Free-flowing, Reagent Grade, anhydrous, 99.9%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
SMILES [Cl-].[Cs+]

Specifications

Melting Point 645°C (literature)
CAS Max % 100.0000%
Quantity 1 kg
Linear Formula CsCl
IUPAC Name cesium;chloride
Formula Weight 168.36
Percent Purity 99.9%
Grade Reagent
Packaging Poly Bottle
Density 3.98 g/cm3
Chemical Name or Material Cesium chloride, Free-flowing, Reagent Grade, anhydrous, 99.9%
Show More Show Less
ShelfLife 720 days from date of manufacture
EINECSNumber 231-600-2
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.