Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Chloride, MP Biomedicals™

Catalog No. MP04813069 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04813069 1 kg
MP04813061 100 g
MP04813063 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP04813069 Supplier MP Biomedicals, Inc Supplier No. 04813069
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cesium Chloride Ultra Pure, 99.999%, White Powder

  • Cesium chloride is typically used for density gradient work and for the purification of virus/phage, nucleic acids and nucleoproteins.
  • It is used for the preparation of electrically conducting glasses, used to make solutions for the separation of RNA from DNA by density gradient centrifugation.
  • Cesium chloride (CsCl) is commonly used in the separation of DNA and RNA.
  • Cesium chloride , an inorganic compound, is an important source of cesium ions in a variety of applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym Cesium Monochloride, Dicesium Dichloride
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C (literature)
Melting Point 646°C (literature)
Color White
pH 5.5 to 6.5 (1% aq soln)
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.999%
Solubility Information Freely soluble in water (500mg/mL),(50% aqueous-clear, colorless solution); soluble in ethanol, alcohol
Formula Weight 168.36 g/mol
Grade 0.99999
Refractive Index 1.642(Lit.)
Appearance White Powder
Density 3.99g/mL(Lit.)
Chemical Name or Material Cesium Chloride
Show More Show Less
storageNote1 Store at Room Temperature (15-30°C), desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.