Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium chloride, MP Biomedicals™

Catalog No. MP04813711 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04813711 100 g
MP04813729 500 g
MP04813700 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP04813711 Supplier MP Biomedicals, Inc Supplier No. 04813711
Add to Cart
Add to Cart

Cesium chloride Optical Grade, 99.9%, White Powder

  • Cesium chloride is used for the preparation of electrically conducting glasses. Used to make solutions for the separation of RNA and DNA, virus/phage by density gradient centrifugation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Melting Point 645°C (literature)
Color White
pH 5.5 to 6.5 (1% aq soln)
Quantity 100 g
Solubility Information Freely soluble in water (50% aqueous - clear, colorless solution); soluble in ethanol, alcohol.
Grade 0.999
Physical Form Powder
RTECSNumber FK9625000
storageNote1 Store at Room Temperature (15-30°C). Store dessicated.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.