Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Chloride, MP Biomedicals™

Catalog No. MP215058980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215058980 100 g
MP215058905 5 g
MP215058925 25 g
MP215058990 500 g
MP215058991 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP215058980 Supplier MP Biomedicals, Inc Supplier No. 0215058980
Add to Cart
Add to Cart

Cesium Chloride, Ultra Pure Reagent, 99.999%, White Powder

  • Cesium chloride is used for the preparation of electrically conducting glasses.
  • It is also used to make solutions for the separation of RNA from DNA by density gradient centrifugation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.