Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Chloride, MP Biomedicals™

Catalog No. MP215058991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215058991 1 kg
MP215058905 5 g
MP215058925 25 g
MP215058980 100 g
MP215058990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP215058991 Supplier MP Biomedicals, Inc Supplier No. 0215058991
Add to Cart
Add to Cart

Cesium Chloride, Ultra Pure Reagent, 99.999%, White Powder

  • Cesium chloride is used for the preparation of electrically conducting glasses.
  • It is also used to make solutions for the separation of RNA from DNA by density gradient centrifugation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C (Literature)
Melting Point 646°C (literature)
Color White
pH 5.5 to 6.5 (1% aq soln)
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.999%
Synonym Cesium Monochloride, Dicesium Dichloride
Solubility Information Very soluble in water. Soluble in ethanol.
Formula Weight 168.35 g/mol
Grade 0.99999
Refractive Index 1.6418 (Lit.)
Appearance White Powder
Density 3.99g/mL (Lit.)
Chemical Name or Material Cesium Chloride
Show More Show Less
RTECSNumber FK9625000
storageNote1 Store at Room Temperature, (15-30°C), desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.