Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cesium Chloride, OmniPur™, Calbiochem™,

Catalog No. 30401KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-401-KG 1 kg
30-301-00GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-401-KG Supplier MilliporeSigma™ Supplier No. 30401KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity cesium chloride for analytical applications

  • Inorganic chemical suitable for laboratory and analytical applications
  • Calbiochem™ molecular agent for the laboratory and manufacturing
  • Nanostructured polyoxometalate-based heterogeneous electrode materials for electrochemical sensing of glucose
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Melting Point 646°C
pH 7 to 9
Quantity 1 kg
Assay Percent Range ≥99.5%
Solubility Information Soluble in water
InChI Formula 1S/ClH.Cs/h1H;/q;+1/p-1
Formula Weight 168.4
Grade Analytical
Product Line EMSURE™
Appearance White Solid
Density 3.97 g/cm³
Chemical Name or Material Cesium Chloride
Show More Show Less
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.