Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Chloride, Optical Grade, 99%, Spectrum™ Chemical
SDP

Catalog No. 18609626 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-626 5 kg Poly Pail
18-609-622 25 g Amber Glass Bottle
18-609-623 100 g Amber Glass Bottle
18-609-624 500 g Amber Glass Bottle
18-609-625 1 kg Amber Glass Bottle
18-609-627 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-609-626 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CE1175KGBL
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7647-17-8

Specifications

CAS Max % 1
Quantity 5 kg
Percent Purity 99%
Grade Optical
Packaging Poly Pail
Appearance Colorless Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.