Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium chloride, Puriss. p.a., ≥99.5%, Solstice

Catalog No. 60045977 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
6 x 100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-977 6 x 100 g
60-023-89 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-977 Supplier Solstice Supplier No. 318076X100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]

Specifications

Melting Point 645°C (literature)
CAS Max % 100.0000%
Quantity 6 x 100 g
Assay Percent Range ≥99.5%
Linear Formula CsCl
UN Number NONH for all modes of transport
Formula Weight 168.36
Percent Purity >99.5%
Grade puriss. p.a.
Packaging Poly Bottle
Product Line Puriss. p.a.
Density 3.98 g/cm3
Chemical Name or Material Cesium chloride, Puriss. p.a., ≥99.5%
Show More Show Less
Hazard Statement P280
Precautionary Statement H361
ShelfLife 1800 days from date of manufacture
EINECSNumber 231-600-2
RTECSNumber FK9625000
Recommended Storage Room Temp
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.