Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Chloride, Sigma-Aldrich

Catalog No. 01258924 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-924 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-924 Supplier MilliporeSigma Supelco Supplier No. C3011500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory chemcial for density gradient centrifugation separation of RNA and DNA

Solutions may contain a slight haze.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula CsCl
Molecular Weight (g/mol) 168.36 g/mol
MDL Number MFCD00010955
Synonym Caesium Chloride, Cesium Monochloride
SMILES [Cl-].[Cs+]

Specifications

Melting Point 645°C
Physical Form Adhering Powder and/or Lumps
Quantity 500 g
InChI Formula 1S/ClH.Cs/h1H;/q;+1/p-1
Formula Weight 168.36 g/mol
Percent Purity ≥99%
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Cesium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.