Learn More
Cesium Chloride, Technical, 98%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
- Separation of various types of DNA by isopycnic centrifugation
- Identification of ions by the color and morphology of the precipitate
Specifications
Specifications
| Melting Point | 645°C |
| Color | White |
| CAS Max % | 1 |
| Quantity | 25 g |
| Assay Percent Range | 98% |
| Molecular Formula | CsCl |
| MDL Number | MFCD00003463 |
| Synonym | Cesium Chloride |
| Solubility Information | Very Soluble in Water |
| Percent Purity | 98% |
| Show More |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.