Learn More
Description
- Separation of various types of DNA by isopycnic centrifugation
- Identification of ions by the color and morphology of the precipitate
Specifications
Specifications
| Boiling Point | 1388°C |
| Melting Point | 645°C |
| Color | Colorless to White |
| CAS Max % | 1 |
| Physical Form | Crystals or Needles |
| Quantity | 5 kg |
| Assay Percent Range | 98% |
| Molecular Formula | CsCl |
| Synonym | Cesium Chloride |
| Solubility Information | Soluble in Water, Methanol, Ethanol |
| Show More |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
