Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium chloride, ultra dry, 99.9% (metals basis)

Catalog No. AA4283222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4283222 100 g
AA4283206 5 g
AA4283214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4283222 Supplier Thermo Scientific Chemicals Supplier No. 04283222
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7647-17-8 | ClCs | 168.36 g/mol

Cesium chloride is widely used in isopycnic centrifugation for separating various types of DNA. It is a reagent in analytical chemistry, where it is used to identify ions by the color and morphology of the precipitate. When enriched in radioisotopes, such as 137CsCl or 131CsCl, cesium chloride is used in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C
Melting Point 646°C
Physical Form Powder
Quantity 100 g
Merck Index 14,2011
Solubility Information Soluble in water (1860g/L) at 20°C,and ethanol. Insoluble in acetone.
IUPAC Name caesium(1+) chloride
Formula Weight 168.36
Percent Purity 99.9%
Odor Odorless
Packaging Ampule
Refractive Index 1.6418
Sensitivity Hygroscopic
Density 3.99 g/mL
Assay (metals basis)
Chemical Name or Material Cesium chloride, Ultra dry
Show More Show Less
Hazard Category H361-H373
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 231-600-2
RTECSNumber FK9625000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.