Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Iodide, Approx. 99.9%, Spectrum™ Chemical
SDP

Catalog No. 18609637 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
5 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-637 100 g Amber Glass Bottle
18-609-639 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-609-637 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1483100GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicative grade chemical suitable for general industrial use and research purpose

  • Suitable for general industrial or research use
  • Used as an input phosphor in x-ray image intensifier tubes for fluoroscopy equipment
  • Supplied by spectrum are indicative of a grade suitable for general industrial use or research purposes
  • Not suitable for human consumption or therapeutic use

Chemical Identifiers

CAS 7789-17-5

Specifications

Boiling Point 685°C
Melting Point 29°C
Color Gold, Silver-gold
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 100 g
Assay Percent Range 99.90%
Molecular Formula CsI
UN Number UN1407
Molecular Weight (g/mol) 259.81
Formula Weight 132.91 g/mol
Percent Purity ∼99.9%
Grade Ungraded
Packaging Amber Glass Bottle
Appearance Solid
Chemical Name or Material Cesium Iodide
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.