Learn More
Cetyl Alcohol, NF, 90-102%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Specifications
Specifications
| CAS Max % | 100% |
| Boiling Point | 344°C (651.2°F) |
| Ignition Residue | 0.1% |
| Packaging | Fiber Drum |
| Quantity | 12 kg |
| Percent Purity | 90-102% |
| Grade | NF |
| Physical Form | Powder or Waxy Solid |
| Chemical Name or Material | Cetyl Alcohol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.