Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetylpyridinium Chloride, Monohydrate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601561 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-561 125 g Glass Bottle
18-601-562 500 g Glass Bottle
18-601-563 2.5 kg Glass Bottle
18-601-565 50 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-561 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CE121125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

CE121, 6004-24-6, C21H38ClN·H2O

Spectrum™ Chemical Cetylpyridinium Chloride, Monohydrate, USP is used as an oral antiseptic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6004-24-6
Molecular Formula C21H40ClNO
Molecular Weight (g/mol) 358.01
MDL Number MFCD00149977
InChI Key NFCRBQADEGXVDL-UHFFFAOYSA-M
IUPAC Name 1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride
SMILES O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Packaging Glass Bottle
Quantity 125 g
UN Number UN2811
Percent Purity 98-102%
Grade USP
Physical Form Powder or Waxy Solid
Chemical Name or Material Cetylpyridinium Chloride, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.