Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetylpyridinium Chloride, MP Biomedicals™

Catalog No. MP219017780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219017780 100 g
MP219017725 25 g
MP219017790 500 g
MP219017791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP219017780 Supplier MP Biomedicals, Inc Supplier No. 0219017780
Add to Cart
Add to Cart

Kills bacteria and other microorganisms. Cetylpyridinium Chloride, MP Biomedicals is an antiseptic that is used in some types of mouthwashes, toothpastes, lozenges, throat sprays, breath sprays and nasal sprays. It has been shown to be effective in preve

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-03-5
Molecular Formula C21H38ClN
Molecular Weight (g/mol) 339.992
InChI Key YMKDRGPMQRFJGP-UHFFFAOYSA-M
Synonym cetylpyridinium chloride, 1-hexadecylpyridinium chloride, hexadecylpyridinium chloride, pristacin, 1-hexadecylpyridin-1-ium chloride, cetamium, dobendan, pyrisept, tserigel, aktivex
PubChem CID 31239
ChEBI CHEBI:32915
IUPAC Name 1-hexadecylpyridin-1-ium;chloride
SMILES CCCCCCCCCCCCCCCC[N+]1=CC=CC=C1.[Cl-]

Specifications

Physical Form Crystalline Powder
Quantity 100 g
Solubility Information Very soluble in water, alcohol and chloroform; Slightly soluble in benzene and ether.
Formula Weight 340g/mol
Recommended Storage Store at Room Temperature (15–30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.