Learn More
ATCC ATCC CF52.Tr (CRL-6244) Canine Adherent Fibroblast Cell Line, BSL-1
Supplier:  ATCC CRL6244
Encompass
CF52.Tr (CRL-6244) is a canine fibroblast adherent cell line from ATCC's NBL Collection supplied for in vitro research applications. The line is maintained under standard mammalian culture conditions and handled at biosafety level 1.
  • Derived from Canis familiaris (dog).
  • Fibroblast cell type with adherent growth properties.
  • Recommended medium: DMEM with 4 mM L-glutamine and 10% fetal bovine serum.
  • Incubation: 37°C in a 5% CO2 atmosphere.
  • Biosafety level 1 handling.
  • Part of the NBL Collection and fully accessioned for research use.
Catalog No. NC3360099