Learn More
Biotium CF647 Maleimide
Supplier:  Biotium 92027
Encompass_Preferred
CF Dye Maleimides are thiol-reactive fluorescent dyes. Maleimides are commonly used to label proteins on free thiol groups. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF647 dye has excitation/emission at 650/665 nm.
Catalog No. 50-196-4847
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.