Learn More
Biotium CF680 Goat Anti-Mouse IgG (H+L), Highly Cross-Adsorbed, single label for STORM, 1 mg/mL
Supplier: Biotium 20817500UL
This product is prepared by labeling highly cross-adsorbed goat anti-mouse IgG (H+L) with CF680 dye. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red/near-IR fluorescent CF680 dye has excitation/emission at 681/698 nm. To minimize crossreactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, hamster, horse, human, rabbit, rat and sheep serum. Single label antibodies have an average of one dye molecule per antibody molecule, which is reported to be optimal for super-resolution imaging by STORM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.