Learn More
Biotium CF750 Free Acid
Supplier:  Biotium 92071
Encompass_Preferred
Near-IR CF dye free acids are control dyes for studies that involve CF dye succinimidyl ester conjugated to antibody, protein or peptides. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Near-infrared fluorescent CF750 dye has excitation/emission at 755/777 nm.
Catalog No. 50-196-4867