Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CFD (Human) ELISA Kit

Catalog No. 89028427 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma, Serum, Urine
Cell Culture, Milk, Plasma, Saliva, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-427 CCS, Plasma, Serum, Urine
89-101-119 Cell Culture, Milk, Plasma, Saliva, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-427 Supplier Abnova™ Supplier No. KA1691

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human CFD

Specifications

Conjugate Biotin
Cross Reactivity Human
Description CFD (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CFD.
Detection Limit 4pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials Detected Polyclonal Antibody, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B,, 2 vials detection Antibody MSP,2 vials Biotinylated Peptide,8μL HRP Streptavidi
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum, Urine
Sample Volume 100 μL
Content And Storage Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type CFD (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.