Learn More
Cytek Biosciences Inc cFluor® V505 anti-Human CD8 (SK1), Quantity 100 Tests
Supplier:  Cytek Biosciences Inc R720255
Encompass
The SK1 monoclonal antibody binds to human CD8a, a 32-34-kDa type I glycoprotein that forms either as an α/α homodimer α/β heterodimer with its isoform CD8b. CD8 is found on thymocytes, a subset of peripheral blood T cells, majority expressing α/β heterodimer, and NK cells, majority expressing α/α homodimer1. CD8 plays a role in antigen recognition and class I MHC-restricted T cell development. CD8 functions are associated with two domains of CD8a. CD8a ectodomain binding to the α3 domain of MHC class I increases the adhesion between CD8+ T lymphocytes and target cells. The cytoplasmic tail binding to the tyrosine kinase p56lck contributes to signal transduction and T cell activation2,3. The antibody was conjugated to a fluorophore and purified by affinity chromatography.
Catalog No. NC2825076
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.