Learn More
Cytek Biosciences Inc cFluor B515 aHu CD141 Anti
Supplier: Cytek Biosciences Inc R720114
Product Name: cFluor B515 Anti-Human CD141 (M80)
Product Class: Antibody
Type: Primary
Product Name: cFluor B515 Anti-Human CD141 (M80)
Cytek Catalog Number: R7-20114
Marker: CD141
Target Species: Human
Clonality: Monoclonal
Clone: M80
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B515
Size: 25 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.