Learn More
Cytek Biosciences Inc cFluor B532 aHu CD3 Anti
Supplier: Cytek Biosciences Inc R721359
Product Name: cFluor B532 Anti-Human CD3 (SK7)
Product Class: Antibody
Type: Primary
Product Name: cFluor B532 Anti-Human CD3 (SK7)
Cytek Catalog Number: R7-21359
Marker: CD3
Target Species: Human
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: SK7
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B532
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.