Learn More
Cytek Biosciences Inc cFluor B532 aHu CD8a Anti
Supplier: Cytek Biosciences Inc R721352
Product Name: cFluor B532 Anti-Human CD8a (RPA-T8)
Product Class: Antibody
Type: Primary
Product Name: cFluor B532 Anti-Human CD8a (RPA-T8)
Cytek Catalog Number: R7-21352
Marker: CD8a
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque|Pigtail Monkey|Sooty Mangabey
Clonality: Monoclonal
Clone: RPA-T8
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B532
Size: 25 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.