Learn More
Cytek Biosciences Inc cFluor B690 aHu CD38 Anti
Supplier: Cytek Biosciences Inc R720299
Product Name: cFluor B690 Anti-Human CD38 (HB-7)
Product Class: Antibody
Type: Primary
Product Name: cFluor B690 Anti-Human CD38 (HB-7)
Cytek Catalog Number: R7-20299
Marker: CD38
Target Species: Human
Clonality: Monoclonal
Clone: HB-7
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B690
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.