Learn More
Cytek Biosciences Inc cFluor BYG667 aMu CD317 Anti
Supplier: Cytek Biosciences Inc R720553
Product Name: cFluor BYG667 Anti-Mouse CD317 (927)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG667 Anti-Mouse CD317 (927)
Cytek Catalog Number: R7-20553
Marker: CD317
Target Species: Mouse
Clonality: Monoclonal
Clone: 927
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor BYG667
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.